Ethyl 3-(3,4-dimethoxyphenyl)-3-oxopropanoate 95%

Supplier: Ambeed
Warning

A207388-250MG A207388-1G
103817-790EA 20 USD
103817-790 103848-470 76873-596 77086-748
Ethyl 3-(3,4-dimethoxyphenyl)-3-oxopropanoate 95%
Ethyl 3-(3,4-dimethoxyphenyl)-3-oxopropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₅
MW: 252.26 g/mol
Boiling Pt: 350.9 °C (760 mm Hg)
Melting Pt: 37…41 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03424826
CAS Number: 4687-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR