Ethyl picolinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-pyridinecarboxylate, Ethyl 2-picolinate

A159061-100G A159061-10G A159061-5G A159061-25G
103843-434EA 5 USD
103843-434 76997-698 76997-700 77300-386 77319-306
Ethyl picolinate 98%
Ethyl picolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 243.7 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00006292
CAS Number: 2524-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR