Ethyl-2-oxocyclopentanecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethylcyclopentanone-2-carboxylate, Ethyl 2-oxocyclopentanecarboxylate

A166152-100G A166152-10G A166152-500G A166152-1000G
103808-966EA 90 USD
103808-966 103844-276 77191-266 77191-268 77191-270 77320-510 77320-512
Ethyl-2-oxocyclopentanecarboxylate 98%
Ethyl-2-oxocyclopentanecarboxylate

Precaución: Sealed in dry, at room temperature.

Formula: C₈H₁₂O₃
Boiling Pt: 228.2 °C (760 mm Hg)
Flash Pt: 77.77 °C
Storage Temperature: Ambient
MDL Number: MFCD00001412
CAS Number: 611-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR