Ethyl-2-oxocyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxocyclohexanecarboxylate

A129608-100G A129608-10G A129608-500G A129608-5G A129608-25G
76996-282EA 59.42 USD
76996-282 76996-284 76996-286 77298-354 77313-688
Ethyl-2-oxocyclohexanecarboxylate 95%
Ethyl-2-oxocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
MW: 170.21 g/mol
Boiling Pt: 230 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00001631
CAS Number: 1655-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR