Ethyl-2-oxobutanoate 92%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-oxobutanoate, Ethyl 2-oxobutyrate

A103146-100G A103146-10G A103146-25G A103146-1G A103146-5G
103796-490EA 7 USD
103796-490 103822-194 77025-254 77025-256 77025-258 77283-046 77283-048
Ethyl-2-oxobutanoate 92%
Ethyl-2-oxobutanoate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
Storage Temperature: Refrigerator
MDL Number: MFCD03265336
CAS Number: 15933-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR