Ethyl-2-methylthiazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-methylthiazole-4-carboxylate
A202401-1G
A202401-25G
A202401-100G
A202401-10G
103817-384EA
8
USD
103817-384
103848-024
77086-136
77086-138
77192-366
77192-368
Ethyl-2-methylthiazole-4-carboxylate 97%
Ethyl 2-methylthiazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉NO₂S Boiling Pt: 242.1 °C (760 mm Hg) Melting Pt: 54…56 °C Flash Pt: 100.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00156148 CAS Number: 6436-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...