Ethyl-2-methylquinoline-6-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methylquinoline-6-carboxylate

A272359-10G A272359-1G
103813-094EA 28 USD
103813-094 103853-004 76801-994 77092-794
Ethyl-2-methylquinoline-6-carboxylate 97%
Ethyl-2-methylquinoline-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.252 g/mol
Boiling Pt: 334.4 °C (760 mm Hg)
Flash Pt: 156.1 °C
Storage Temperature: Ambient
MDL Number: MFCD09879697
CAS Number: 855763-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR