4-Carbethoxy-3-methyl-2-cyclohexen-1-one 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methyl-4-oxocyclohex-2-enecarboxylate

A619119-1G A619119-5G A619119-100G A619119-10G A619119-500G A619119-25G
103840-204EA 14 USD
103840-204 76844-376 76844-378 77292-912 77292-914 77292-916 77307-958
4-Carbethoxy-3-methyl-2-cyclohexen-1-one 97%
4-Carbethoxy-3-methyl-2-cyclohexen-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MW: 182.216 g/mol
Boiling Pt: 271.8 °C (760 mm Hg)
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001582
CAS Number: 487-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR