Ethyl-2-methyl-1H-pyrrole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methyl-1H-pyrrole-3-carboxylate

A227075-1G A227075-25G A227075-5G A227075-250MG A227075-10G
103819-292EA 12 USD
103819-292 103850-200 77088-950 77088-952 77088-954 77261-466 77748-375
Ethyl-2-methyl-1H-pyrrole-3-carboxylate 98%
Ethyl-2-methyl-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.1784 g/mol
Boiling Pt: 289.7 °C (760 mm Hg)
Flash Pt: 129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00101795
CAS Number: 936-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR