Ethyl-2-methoxy-6-methylbenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-methoxy-6-methylbenzoate, 2-Methoxy-6-methylbenzoic Acid Ethyl Ester
A159025-10G
A159025-250MG
A159025-1G
A159025-5G
103805-214EA
7
USD
103805-214
103843-426
77259-526
77259-528
77319-288
77319-290
Ethyl-2-methoxy-6-methylbenzoate 97%
Ethyl-2-methoxy-6-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00191666 CAS Number: 6520-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...