Ethyl 2-hydroxypyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

A405115-1G A405115-100MG A405115-250MG A405115-5G
103827-114EA 42 USD
103827-114 103838-156 76841-644 77033-704 77033-706 77307-096
Ethyl 2-hydroxypyrimidine-5-carboxylate 97%
Ethyl 2-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991546
CAS Number: 95928-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR