Ethyl glycolate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl Hydroxyacetate, Hydroxyacetic Acid Ethyl Ester, Glycolic Acid Ethyl Ester, Ethyl 2-hydroxyacetate
A305055-100G
A305055-10G
A305055-1G
A305055-500G
A305055-5G
A305055-1000G
A305055-25G
103810-040EA
108
USD
103810-040
76802-604
76802-606
76802-608
76802-610
76802-612
76838-290
77305-906
Ethyl glycolate 97%
Ethyl glycolate
Precaución: for Research Use Only
Formula:
C₄H₈O₃ Boiling Pt: 158…159 °C (760 mm Hg) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021970 CAS Number: 623-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...