Ethyl-2-hydrazinylacetate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl hydrazinoacetate hydrochloride, Ethyl 2-hydrazinylacetate Hydrochloride, Ethyl hydrazinoacetate HCl, Ethyl 2-hydrazinylacetate hydrochloride
A131610-100G
A131610-10G
A131610-500G
A131610-1G
A131610-25G
A131610-5G
103801-610EA
8
USD
103801-610
77136-926
77136-928
77136-930
77298-498
77298-500
77314-112
Ethyl-2-hydrazinylacetate hydrochloride 97%
Ethyl-2-hydrazinylacetate hydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₁ClN₂O₂ MW: 154.594 g/mol Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012923 CAS Number: 6945-92-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...