Ethyl-2-formyl-3-oxopropionate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-formyl-3-oxopropanoate, Ethyl 2-Formyl-3-oxopropionate, Diformylacetic acid ethyl ester, 2-Formyl-3-oxopropionic acid ethyl ester, Ethyl diformylacetate

A460904-100G A460904-10G A460904-25G A460904-5G A460904-1G
103827-918EA 9 USD
103827-918 103838-996 76789-476 76789-478 76842-874 76842-876 77307-424
Ethyl-2-formyl-3-oxopropionate 97%
Ethyl-2-formyl-3-oxopropionate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 181.5 °C (760 mm Hg)
Flash Pt: 67.423 °C
Storage Temperature: Freezer
MDL Number: MFCD11112084
CAS Number: 80370-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR