Ethyl-2-fluoropropionate 97%

Supplier: Ambeed
Danger

Synonyms: 2-Fluoropropionic Acid Ethyl Ester, Ethyl 2-fluoropropionate

A101862-25G A101862-1G A101862-5G
103796-182EA 12 USD
103796-182 103821-878 76796-292 77308-768 77308-770
Ethyl-2-fluoropropionate 97%
Ethyl-2-fluoropropionate

Precaución: for Research Use Only

Formula: C₅H₉FO₂
Boiling Pt: 125 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01320801
CAS Number: 349-43-9
UN: 3272
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR