Ethyl-2-ethylacetoacetate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-ethyl-3-oxobutanoate

A374861-100G A374861-10G A374861-500G A374861-5G
103837-582EA 9 USD
103837-582 76773-492 76773-494 76773-496 77306-864
Ethyl-2-ethylacetoacetate 97%
Ethyl-2-ethylacetoacetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2
Boiling Pt: 190 °C (760 mm Hg)
Density: 0.9937
Flash Pt: 69 °C
Storage Temperature: Ambient
MDL Number: MFCD00039898
CAS Number: 607-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR