Ethyl 2-chloroquinazoline-4-carboxylate 97%

Supplier: Ambeed
Warning

A148403-100MG A148403-5G A148403-250MG
103803-926EA 226 USD
103803-926 103842-038 77027-986 77299-652 77667-108
Ethyl 2-chloroquinazoline-4-carboxylate 97%
Ethyl 2-chloroquinazoline-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉ClN₂O₂
Boiling Pt: 300.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11518957
CAS Number: 1092352-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR