Ethyl-2-chloropyrimidine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloropyrimidine-5-carboxylate

A173615-100G A173615-100MG A173615-10G A173615-250MG A173615-25G A173615-5G A173615-1G
103806-844EA 8 USD
103806-844 103845-144 76784-318 76784-320 76784-322 76784-324 77082-400 77082-402 77301-288
Ethyl-2-chloropyrimidine-5-carboxylate 98%
Ethyl-2-chloropyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.60 g/mol
Boiling Pt: 314.2 °C (760 mm Hg)
Flash Pt: 143.816 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09863164
CAS Number: 89793-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR