Ethyl 2-chloropyrimidine-4-carboxylate 95%

Supplier: Ambeed
Warning

A204018-1G A204018-250MG
103848-180EA 361 USD
103848-180 76814-388 76814-390
Ethyl 2-chloropyrimidine-4-carboxylate 95%
Ethyl 2-chloropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₂
MW: 186.6 g/mol
Boiling Pt: 314.2±15.0 °C (760 mm Hg)
Flash Pt: 143.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275109
CAS Number: 1196152-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR