Ethyl-2-chloro-4-oxazolecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chlorooxazole-4-carboxylate

A148919-250MG A148919-1G A148919-5G
103803-978EA 19 USD
103803-978 103842-098 77162-668 77317-566 77317-568
Ethyl-2-chloro-4-oxazolecarboxylate 97%
Ethyl 2-chloro-4-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₆ClNO₃
MW: 175.57 g/mol
Melting Pt: 52…56 °C
Storage Temperature: Freezer
MDL Number: MFCD06660120
CAS Number: 460081-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR