Ethyl-2-chloro-5-nitropyridine-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-5-nitroisonicotinate

A716293-25G A716293-1G A716293-5G A716293-10G
103840-710EA 63 USD
103840-710 76651-462 77308-184 77308-186 77783-743
Ethyl-2-chloro-5-nitropyridine-4-carboxylate 97%
Ethyl-2-chloro-5-nitropyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₄
MW: 230.61 g/mol
Melting Pt: 40…41 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22126101
CAS Number: 907545-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR