Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-4-methylpyrimidine-5-carboxylate

A303633-100MG A303633-250MG A303633-1G A303633-25G A303633-5G
103814-384EA 41 USD
103814-384 103835-550 77032-502 77032-504 77305-876 77305-878 77305-880
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate 97%
Ethyl-2-chloro-4-methylpyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.622 g/mol
Boiling Pt: 302.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03425963
CAS Number: 188781-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR