Ethyl-2-chloro-2-(hydroxyimino)acetate 97%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-2-(hydroxyimino)acetic acid ethyl ester, alpha-Chloroglyoxalic acid ethyl ester Oxime, Ethylchloroximidoacetate, Ethyl 2-Chloro-2-(hydroxyimino)acetate

A123916-10G A123916-25G A123916-1G A123916-5G
103800-462EA 5 USD
103800-462 76870-390 76870-392 77312-586 77312-588
Ethyl-2-chloro-2-(hydroxyimino)acetate 97%
Ethyl-2-chloro-2-(hydroxyimino)acetate

Precaución: for Research Use Only

Formula: C₄H₆ClNO₃
Melting Pt: 70…76 °C
Flash Pt: 93.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010209
CAS Number: 14337-43-0
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR