2-Butynoic acid ethyl ester 97%

Supplier: Ambeed
Warning

A188953-10G A188953-500G A188953-1G A188953-5G A188953-25G A188953-100G
103816-148EA 5 USD
103816-148 103846-742 76769-822 76769-824 77084-488 77084-490 77302-162 77625-904
2-Butynoic acid ethyl ester 97%
2-Butynoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₆H₈O₂
Boiling Pt: 163 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015182
CAS Number: 4341-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR