Ethyl 2-bromothiophene-3-carboxylate 95%

Supplier: Ambeed
Warning

A117992-10G A117992-250MG A117992-25G A117992-1G A117992-5G
103798-972EA 17 USD
103798-972 103824-862 77026-234 77026-236 77026-238 77297-570 77297-572
Ethyl 2-bromothiophene-3-carboxylate 95%
Ethyl 2-bromothiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇BrO₂S
Boiling Pt: 257.7±20.0 °C (760 mm Hg)
Flash Pt: 109.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09965551
CAS Number: 632325-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR