Ethyl-2-bromothiazole-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromothiazole-5-carboxylate

A392431-1G A392431-25G A392431-5G A392431-100G A392431-10G A392431-250MG
103826-902EA 6 USD
103826-902 103837-936 76841-334 76841-336 76841-338 77115-658 77115-660 77686-086
Ethyl-2-bromothiazole-5-carboxylate 97%
Ethyl-2-bromothiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.08 g/mol
Boiling Pt: 277.6 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00463837
CAS Number: 41731-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR