Ethyl-2-bromothiazole-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A241927-100G
A241927-10G
A241927-250MG
A241927-500G
A241927-1G
A241927-25G
A241927-5G
103811-442EA
6
USD
103811-442
103851-250
76801-356
76801-358
76801-360
76801-362
77090-410
77090-412
77090-414
Ethyl-2-bromothiazole-4-carboxylate 97%
Ethyl-2-bromothiazole-4-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₆BrNO₂S Boiling Pt: 277.6 °C (760 mm Hg) Melting Pt: 48…52 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788564 CAS Number: 100367-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...