Ethyl-2-bromo-4-oxazolecarboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-bromooxazole-4-carboxylate
A162151-10G
A162151-250MG
A162151-25G
A162151-5G
103805-578EA
14
USD
103805-578
103843-828
76783-906
76783-908
77319-862
77319-864
Ethyl-2-bromo-4-oxazolecarboxylate 97%
Ethyl 2-bromo-4-oxazolecarboxylate
Precaución: for Research Use Only
Formula:
C₆H₆BrNO₃ MW: 220.0207 g/mol Boiling Pt: 272 °C (760 mm Hg) Flash Pt: 118 °C Storage Temperature: Freezer |
MDL Number:
MFCD09801023 CAS Number: 460081-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...