Ethyl-2-bromo-6-benzothiazolecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromo-6-benzothiazolecarboxylate

A123743-250MG A123743-1G A123743-5G
103800-436EA 17 USD
103800-436 77258-156 77312-556 77312-558
Ethyl-2-bromo-6-benzothiazolecarboxylate 97%
Ethyl-2-bromo-6-benzothiazolecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈BrNO₂S
MW: 286.1500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669850
CAS Number: 99073-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR