Ethyl-2-bromo-5-nitrobenzoate 98%
Supplier: Ambeed
|
Warning
|
A221028-10G
A221028-1G
A221028-25G
A221028-5G
103818-884EA
25
USD
103818-884
103849-764
76975-828
77303-570
77303-572
77303-574
Ethyl-2-bromo-5-nitrobenzoate 98%
Ethyl-2-bromo-5-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrNO₄ Boiling Pt: 339.3 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD12025203 CAS Number: 208176-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...