Ethyl 2-bromo-4-methyl-1H-imidazole-5-carboxylate 97%

Supplier: Ambeed
Warning

A315640-100MG A315640-250MG A315640-1G
103825-080EA 134 USD
103825-080 103836-002 76816-944 76816-946 77306-134
Ethyl 2-bromo-4-methyl-1H-imidazole-5-carboxylate 97%
Ethyl 2-bromo-4-methyl-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O₂
MW: 233.06 g/mol
Boiling Pt: 265.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09033863
CAS Number: 95470-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR