Ethyl-2-amino-5-oxazolecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-aminooxazole-5-carboxylate

A135089-10G A135089-250MG A135089-1G A135089-25G
103802-090EA 23 USD
103802-090 77258-594 77258-596 77314-876 77314-878
Ethyl-2-amino-5-oxazolecarboxylate 97%
Ethyl 2-amino-5-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.1393 g/mol
Boiling Pt: 282.9 °C (760 mm Hg)
Melting Pt: 151…153 °C
Flash Pt: 124.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06657433
CAS Number: 113853-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR