Ethyl-2-amino-4-oxazolecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-aminooxazole-4-carboxylate

A104992-100G A104992-10G A104992-250MG A104992-25G A104992-1G A104992-5G
103796-812EA 10 USD
103796-812 103822-540 77008-542 77008-544 77008-546 77008-548 77309-406 77309-408
Ethyl-2-amino-4-oxazolecarboxylate 98%
Ethyl 2-amino-4-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Boiling Pt: 269.1 °C (760 mm Hg)
Melting Pt: 119…121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659907
CAS Number: 177760-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR