Ethyl-2-aminoisonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

A323319-1G A323319-25G A323319-5G A323319-100G A323319-10G A323319-250MG
103825-330EA 6 USD
103825-330 103836-266 76839-182 76839-184 76839-186 77015-288 77015-290 77015-292
Ethyl-2-aminoisonicotinate 98%
Ethyl 2-aminoisonicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD03791260
CAS Number: 13362-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR