Ethyl 2-amino-6-chlorobenzoate 98%

Supplier: Ambeed
Warning

A295181-10G A295181-1G A295181-5G
103814-048EA 27 USD
103814-048 103835-184 76802-438 76837-836 76837-838
Ethyl 2-amino-6-chlorobenzoate 98%
Ethyl 2-amino-6-chlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275014
CAS Number: 172217-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR