Ethyl 2-amino-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

A373139-250MG A373139-1G
103826-526EA 7 USD
103826-526 77101-612 77306-850
Ethyl 2-amino-5-nitrobenzoate 95%
Ethyl 2-amino-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.18 g/mol
Boiling Pt: 385 °C (760 mm Hg)
Flash Pt: 186.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11174616
CAS Number: 32203-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR