Ethyl-2-amino-5-iodobenzoate 98%

Supplier: Ambeed
Warning

A102612-100G A102612-25G A102612-1G
103822-092EA 7 USD
103822-092 76994-578 76994-580 77880-833
Ethyl-2-amino-5-iodobenzoate 98%
Ethyl-2-amino-5-iodobenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀INO₂
Boiling Pt: 347.9 °C (760 mm Hg)
Melting Pt: 69…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04038940
CAS Number: 268568-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR