Ethyl-2-amino-5-bromothiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-5-bromothiazole-4-carboxylate

A248598-5G A248598-10G A248598-25G
103851-664EA 15 USD
103851-664 77091-014 77113-392 77304-450
Ethyl-2-amino-5-bromothiazole-4-carboxylate 97%
Ethyl-2-amino-5-bromothiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O₂S
Boiling Pt: 357 °C (760 mm Hg)
Flash Pt: 169.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06669909
CAS Number: 61830-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR