Ethyl-2-amino-5-bromonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-5-bromonicotinate

A101270-10G A101270-1G A101270-5G
103795-886EA 17 USD
103795-886 103821-572 76994-496 77297-038 77297-040
Ethyl-2-amino-5-bromonicotinate 97%
Ethyl 2-amino-5-bromonicotinate

Precaución: for Research Use Only

Formula: C₈H₉N₂O₂Br
MW: 245.0700 g/mol
Melting Pt: 139…141 °C
Storage Temperature: Ambient
MDL Number: MFCD08276943
CAS Number: 433226-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR