Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate

A200485-1G A200485-5G A200485-250MG
103817-206EA 15 USD
103817-206 103847-834 77085-892 77085-894 77248-838
Ethyl-2-amino-4-phenylthiophene-3-carboxylate 96%
Ethyl-2-amino-4-phenylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂S
MW: 247.32 g/mol
Boiling Pt: 391 °C (760 mm Hg)
Flash Pt: 190.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00126391
CAS Number: 4815-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR