Ethyl-2-amino-4-methyl-5-oxazolecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4-methyloxazole-5-carboxylate

A214323-100MG A214323-250MG A214323-1G A214323-5G
103818-392EA 64 USD
103818-392 103849-098 76873-764 76873-766 77303-338 77303-340
Ethyl-2-amino-4-methyl-5-oxazolecarboxylate 98%
Ethyl 2-amino-4-methyl-5-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₃
MW: 170.1659 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09028430
CAS Number: 79221-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR