Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate

A107500-10G A107500-250MG A107500-25G A107500-1G A107500-5G
103797-200EA 18 USD
103797-200 103822-968 77188-876 77188-878 77188-880 77309-824 77309-826
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₃
MW: 183.17 g/mol
Boiling Pt: 379 °C (760 mm Hg)
Melting Pt: 300 °C
Flash Pt: 183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039708
CAS Number: 15400-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR