Ethyl thiooxamate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Thiooxamic acid ethyl ester, Ethyl amino(thioxo)acetate, Ethyl 2-amino-2-thioxoacetate

A148941-100G A148941-10G A148941-1G A148941-5G A148941-25G A148941-500G A148941-1000G
103803-988EA 5 USD
103803-988 103842-108 76798-542 76798-544 77299-694 77299-696 77317-582 77592-806 77667-124
Ethyl thiooxamate 98%
Ethyl thiooxamate

Precaución: for Research Use Only

Formula: C₄H₇NO₂S
Boiling Pt: 199.2 °C (760 mm Hg)
Melting Pt: 62…65 °C
Flash Pt: 74.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00074903
CAS Number: 16982-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR