2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-amino-1-methyl-1H-imidazole-5-carboxylate
A229412-100MG
A229412-250MG
A229412-5G
103819-476EA
59
USD
103819-476
103850-376
76987-280
76987-282
77089-222
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester 97%
2-Amino-3-methyl-3H-imidazole-4-carboxylic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₇H₁₁N₃O₂ MW: 169.181 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09997799 CAS Number: 177760-04-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...