Ethyl-2,6-dichloropyridine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2,6-Dichloropyridine-3-carboxylate, 2,6-Dichloropyridine-3-carboxylic acid ethyl ester, 2,6-Dichloronicotinic acid ethyl ester, Ethyl 2,6-Dichloronicotinate

A417787-1G A417787-25G A417787-5G A417787-100G A417787-10G A417787-250MG
103827-342EA 8 USD
103827-342 103838-388 76841-980 76841-982 76841-984 77252-308 77252-310 77252-312
Ethyl-2,6-dichloropyridine-3-carboxylate 95%
Ethyl-2,6-dichloropyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.06 g/mol
Boiling Pt: 286 °C (760 mm Hg)
Melting Pt: 45…49 °C
Flash Pt: 126.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09757510
CAS Number: 58584-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR