Ethyl 2,4-dioxohexanoate 97%
Supplier: Ambeed
|
Warning
|
A247981-100MG
A247981-250MG
A247981-5G
103811-804EA
30
USD
103811-804
103851-640
76801-504
76801-506
77304-434
Ethyl 2,4-dioxohexanoate 97%
Ethyl 2,4-dioxohexanoate
Precaución: for Research Use Only
Formula:
C₈H₁₂O₄ MW: 172.18 g/mol Boiling Pt: 233.8 °C (760 mm Hg) Flash Pt: 95.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03265450 CAS Number: 13246-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...