Ethyl 2,4-dihydroxy-6-pentylbenzoate 95%

Supplier: Ambeed
Warning

A379338-25G A379338-5G A379338-10G A379338-1G
103837-680EA 163 USD
103837-680 77290-972 77290-974 77686-060 77686-061
Ethyl 2,4-dihydroxy-6-pentylbenzoate 95%
Ethyl 2,4-dihydroxy-6-pentylbenzoate

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₄
MW: 252.306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11618127
CAS Number: 38862-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR