Ethyl 2,2-dimethyl-1,3-dioxane-5-carboxylate 95%

Supplier: Ambeed
Warning

A187179-10G A187179-1G A187179-25G
103815-994EA 14 USD
103815-994 103846-586 76813-780 77302-062 77567-740
Ethyl 2,2-dimethyl-1,3-dioxane-5-carboxylate 95%
Ethyl 2,2-dimethyl-1,3-dioxane-5-carboxylate

Precaución: For research use only.

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 217.5±25.0 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11975772
CAS Number: 82962-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR