(1-Ethoxycarbonylethylidene)triphenylphosphorane 97% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (Carbethoxyethylidene)triphenylphosphorane, Ethyl 2-(triphenylphosphoranylidene)propanoate

A734502-5G A734502-100G
103810-548EA 347 USD
103810-548 103840-804 77308-232 77552-262
(1-Ethoxycarbonylethylidene)triphenylphosphorane 97% (by HPLC)
(1-Ethoxycarbonylethylidene)triphenylphosphorane

Precaución: for Research Use Only

Formula: C₂₃H₂₃O₂P
MW: 362.41 g/mol
Boiling Pt: 497.2 °C (760 mm Hg)
Melting Pt: 154…160 °C
Flash Pt: 267.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00009160
CAS Number: 5717-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR