Ethyl tetrahydropyran-4-ylacetate 98%

Supplier: Ambeed
Warning

A119655-100MG A119655-250MG A119655-1G A119655-5G A119655-10G
103799-812EA 12 USD
103799-812 76797-278 76797-280 77311-920 77311-922 77666-712
Ethyl tetrahydropyran-4-ylacetate 98%
Ethyl tetrahydropyran-4-ylacetate

Precaución: for Research Use Only

Formula: C₉H₁₆O₃
Boiling Pt: 228.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03788560
CAS Number: 103260-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR